Search
Search

RpS20| Gene Symbol: RpS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079697.3 | NP_524421.1 | 3 - 4 | 344 | 74 | |
| GenBank mRNA | AY071742.1 | - | 3 - 4 | 345 | 74 | |
| KX532014.1 | - | 2 - 3 | 254 | 74 | ||
| Y11119.1 | - | 334 | 74 | |||
Set Membership: |
|