Search
Search

srprb| Gene Symbol: srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002572.1 | NP_001002572.1 | 5 - 6 | 556 | 107 | |
| GenBank mRNA | BC076224.1 | - | 5 - 6 | 556 | 107 | |
| BC095560.1 | - | 5 - 6 | 552 | 107 | ||
| BC164607.1 | - | 5 - 6 | 561 | 107 | ||
Set Membership: |
|