Search
Search

thrap3b| Gene Symbol: thrap3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214815.1 | NP_999980.1 | 8 - 9 | 2212 | 138 | |
| XM_005173742.4 | XP_005173799.1 | 9 - 10 | 2191 | 138 | ||
| XM_021468043.1 | XP_021323718.1 | 8 - 9 | 2324 | 138 | ||
| XM_021468044.1 | XP_021323719.1 | 6 - 7 | 1252 | 138 | ||
| XM_021468045.1 | XP_021323720.1 | 8 - 9 | 1500 | 138 | ||
| GenBank mRNA | BC070004.1 | - | 8 - 9 | 2212 | 138 | |
Set Membership: |
|