Search
Search

tectb| Gene Symbol: tectb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008594.1 | NP_001008594.1 | 4 - 5 | 539 | 109 | |
| XM_017358448.2 | XP_017213937.1 | 4 - 5 | 694 | 109 | ||
| XM_017358450.2 | XP_017213939.1 | 4 - 5 | 693 | 109 | ||
| GenBank mRNA | BC086745.1 | - | 4 - 5 | 539 | 109 | |
| BC164139.1 | - | 3 - 4 | 449 | 109 | ||
| FJ374270.1 | - | 4 - 5 | 539 | 109 | ||
Set Membership: |
|