Search
Search

six3a| Gene Symbol: six3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131362.1 | NP_571437.1 | 1 - 2 | 813 | 55 | |
| GenBank mRNA | AF030280.1 | - | 1 - 2 | 813 | 55 | |
| BC059414.1 | - | 1 - 2 | 804 | 55 | ||
Set Membership: |
|