Search
Search

enpp6| Gene Symbol: enpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013527.1 | NP_001013545.1 | 6 - 7 | 1126 | 97 | |
| GenBank mRNA | BC090903.1 | - | 6 - 7 | 1126 | 97 | |
| BC165573.1 | - | 6 - 7 | 1027 | 97 | ||
Set Membership: |
|