Search
Search

cep70| Gene Symbol: cep70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114576.1 | NP_001108048.1 | 9 - 10 | 910 | 64 | |
| XM_017353091.2 | XP_017208580.1 | 8 - 9 | 64 | |||
| XM_021469287.1 | XP_021324962.1 | 8 - 9 | 64 | |||
| XM_021469288.1 | XP_021324963.1 | 9 - 10 | 64 | |||
| GenBank mRNA | BC154450.1 | - | 9 - 10 | 910 | 64 | |
Set Membership: |
|