Search
Search

PUS10| Gene Symbol: PUS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346586.1 | NP_001333515.1 | 7 - 8 | 1234 | 73 | |
| XM_014731154.2 | XP_014586640.1 | 716 | 73 | |||
| XM_023618399.1 | XP_023474167.1 | 1048 | 73 | |||
| XM_023618400.1 | XP_023474168.1 | 681 | 73 | |||
| XM_023618401.1 | XP_023474169.1 | 765 | 73 | |||
Set Membership: |
|