Search
Search

CD151| Gene Symbol: CD151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005598401.3 | XP_005598458.1 | 606 | 71 | ||
| XM_005598402.3 | XP_005598459.1 | 562 | 71 | |||
| XM_023654800.1 | XP_023510568.1 | 540 | 71 | |||
| XM_023654801.1 | XP_023510569.1 | 535 | 71 | |||
| XM_023654802.1 | XP_023510570.1 | 495 | 71 | |||
| XM_023654803.1 | XP_023510571.1 | 602 | 71 | |||