Search
Search

PPP2R3C| Gene Symbol: PPP2R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623974.1 | XP_023479742.1 | 888 | 73 | ||
| XM_023623985.1 | XP_023479753.1 | 815 | 73 | |||
| XM_023623989.1 | XP_023479757.1 | 720 | 73 | |||
Set Membership: |
|