Search
Search

MAP2K5| Gene Symbol: MAP2K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496608.5 | XP_001496658.1 | 1480 | 82 | ||
| XM_001496625.5 | XP_001496675.1 | 1480 | 82 | |||
| XM_005602976.3 | XP_005603033.1 | 1480 | 82 | |||
| XM_014733752.2 | XP_014589238.1 | 1484 | 82 | |||
| XM_014733753.2 | XP_014589239.1 | 1487 | 82 | |||
| XM_014733754.2 | XP_014589240.1 | 1486 | 82 | |||
| XM_014733755.2 | XP_014589241.1 | 1488 | 82 | |||
| XM_014733756.2 | XP_014589242.1 | 1480 | 82 | |||
| XM_023654936.1 | XP_023510704.1 | 1487 | 82 | |||
| XM_023654942.1 | XP_023510710.1 | 1480 | 82 | |||
| XM_023654948.1 | XP_023510716.1 | 1488 | 82 | |||
| XM_023654950.1 | XP_023510718.1 | 1480 | 82 | |||
| XM_023654952.1 | XP_023510720.1 | 1484 | 82 | |||
| XM_023654953.1 | XP_023510721.1 | 1480 | 82 | |||
| XM_023654955.1 | XP_023510723.1 | 1481 | 82 | |||
| XM_023654958.1 | XP_023510726.1 | 1481 | 82 | |||
| XM_023654960.1 | XP_023510728.1 | 1480 | 82 | |||
| XM_023654961.1 | XP_023510729.1 | 1483 | 82 | |||
| XM_023654964.1 | XP_023510732.1 | 1480 | 82 | |||
| XM_023654969.1 | XP_023510737.1 | 1480 | 82 | |||
| XM_023654971.1 | XP_023510739.1 | 1480 | 82 | |||
| XM_023654972.1 | XP_023510740.1 | 1480 | 82 | |||
Set Membership: |
|