Search
Search

KMT2D| Gene Symbol: KMT2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611139.3 | XP_005611196.2 | 17648 | 63 | ||
| XM_023643474.1 | XP_023499242.1 | 15640 | 63 | |||
| XM_023643475.1 | XP_023499243.1 | 15637 | 63 | |||
| XM_023643476.1 | XP_023499244.1 | 15619 | 63 | |||
| XM_023643477.1 | XP_023499245.1 | 15572 | 63 | |||
| XM_023643478.1 | XP_023499246.1 | 15547 | 63 | |||
| XM_023643479.1 | XP_023499247.1 | 15451 | 63 | |||
Set Membership: |
|