Search
Search

TMEM161B| Gene Symbol: TMEM161B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618029.1 | XP_023473797.1 | 1050 | 66 | ||
| XM_023618030.1 | XP_023473798.1 | 943 | 66 | |||
| XM_023618031.1 | XP_023473799.1 | 6112 | 66 | |||
| XM_023618032.1 | XP_023473800.1 | 925 | 66 | |||
| XM_023618033.1 | XP_023473801.1 | 570 | 66 | |||
Set Membership: |
|