Search
Search

DNPEP| Gene Symbol: DNPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610628.3 | XP_005610685.1 | 553 | 115 | ||
| XM_005610629.3 | XP_005610686.1 | 390 | 115 | |||
| XM_005610630.3 | XP_005610687.1 | 463 | 115 | |||
| XM_005610631.3 | XP_005610688.1 | 432 | 115 | |||
| XM_023642395.1 | XP_023498163.1 | 433 | 115 | |||
Set Membership: |
|