Search
Search

ANKRA2| Gene Symbol: ANKRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023618197.1 | XP_023473965.1 | 1301 | 78 | ||
| XM_023618198.1 | XP_023473966.1 | 1179 | 78 | |||
| XM_023618199.1 | XP_023473967.1 | 1179 | 78 | |||
| XM_023618200.1 | XP_023473968.1 | 1119 | 78 | |||
Set Membership: |
|