Search
Search

MTMR14| Gene Symbol: MTMR14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001915139.5 | XP_001915174.4 | 1177 | 117 | ||
| XM_023620021.1 | XP_023475789.1 | 1177 | 117 | |||
| XM_023620022.1 | XP_023475790.1 | 1177 | 117 | |||
| XM_023620023.1 | XP_023475791.1 | 1177 | 117 | |||
| XM_023620024.1 | XP_023475792.1 | 1177 | 117 | |||
Set Membership: |
|