Search
Search

LMCD1| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001491739.6 | XP_001491789.3 | 1068 | 75 | ||
| XM_005600407.3 | XP_005600464.1 | 2303 | 75 | |||
| XM_005600408.3 | XP_005600465.1 | 973 | 75 | |||
Set Membership: |
|