Search
Search

GADL1| Gene Symbol: GADL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005600868.2 | XP_005600925.1 | 776 | 104 | ||
| XM_005600869.2 | XP_005600926.1 | 695 | 104 | |||
| XM_005600870.3 | XP_005600927.1 | 450 | 104 | |||
Set Membership: |
|