Search
Search

CCDC141| Gene Symbol: CCDC141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001501039.4 | XP_001501089.1 | 2919 | 70 | ||
| XM_023622121.1 | XP_023477889.1 | 2938 | 70 | |||
| XM_023622123.1 | XP_023477891.1 | 2938 | 70 | |||
| XM_023622124.1 | XP_023477892.1 | 2821 | 70 | |||
| XM_023622125.1 | XP_023477893.1 | 2818 | 70 | |||
Set Membership: |
|