Search
Search

ABCC5| Gene Symbol: ABCC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497097.6 | XP_001497147.2 | 4137 | 79 | ||
| XM_023623399.1 | XP_023479167.1 | 3900 | 79 | |||
| XM_023623400.1 | XP_023479168.1 | 3879 | 79 | |||
| XM_023623401.1 | XP_023479169.1 | 7834 | 79 | |||
| XM_023623402.1 | XP_023479170.1 | 3050 | 79 | |||
Set Membership: |
|