Search
Search
| Gene Symbol: CBLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023623831.1 | XP_023479599.1 | 1001 | 65 | ||
| XM_023623832.1 | XP_023479600.1 | 1001 | 65 | |||
| XM_023623833.1 | XP_023479601.1 | 1152 | 65 | |||
| XM_023623834.1 | XP_023479602.1 | 3546 | 65 | |||
Set Membership: |
|