Search
Search

MROH7| Gene Symbol: MROH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607025.3 | XP_005607082.2 | 3064 | 89 | ||
| XM_023629276.1 | XP_023485044.1 | 3146 | 89 | |||
| XM_023629281.1 | XP_023485049.1 | 2990 | 89 | |||
| XM_023629286.1 | XP_023485054.1 | 2968 | 89 | |||
| XM_023629290.1 | XP_023485058.1 | 3064 | 89 | |||
| XM_023629297.1 | XP_023485065.1 | 1577 | 89 | |||
| XM_023629302.1 | XP_023485070.1 | 1711 | 89 | |||
Set Membership: |
|