Search
Search

BEND6| Gene Symbol: BEND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001499638.4 | XP_001499688.1 | 351 | 71 | ||
| XM_014734514.2 | XP_014590000.1 | 418 | 71 | |||
| XM_023624313.1 | XP_023480081.1 | 235 | 71 | |||
| XM_023624314.1 | XP_023480082.1 | 400 | 71 | |||
| XM_023624315.1 | XP_023480083.1 | 435 | 71 | |||
| XM_023624316.1 | XP_023480084.1 | 432 | 71 | |||
Set Membership: |
|