Search
Search

PDZD2| Gene Symbol: PDZD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604325.3 | XP_005604382.1 | 2213 | 97 | ||
| XM_014734921.2 | XP_014590407.1 | 3325 | 97 | |||
| XM_023625798.1 | XP_023481566.1 | 3272 | 97 | |||
| XM_023625799.1 | XP_023481567.1 | 3116 | 97 | |||
| XM_023625800.1 | XP_023481568.1 | 2996 | 97 | |||
| XM_023625801.1 | XP_023481569.1 | 2258 | 97 | |||
| XM_023625802.1 | XP_023481570.1 | 2840 | 97 | |||
| XM_023625803.1 | XP_023481571.1 | 3456 | 97 | |||
Set Membership: |
|