Search
Search

KIF21A| Gene Symbol: KIF21A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643349.1 | XP_023499117.1 | 422 | 93 | ||
| XM_023643350.1 | XP_023499118.1 | 422 | 93 | |||
| XM_023643351.1 | XP_023499119.1 | 422 | 93 | |||
| XM_023643352.1 | XP_023499120.1 | 422 | 93 | |||
| XM_023643353.1 | XP_023499121.1 | 422 | 93 | |||
| XM_023643354.1 | XP_023499122.1 | 437 | 93 | |||
| XM_023643355.1 | XP_023499123.1 | 422 | 93 | |||
| XM_023643356.1 | XP_023499124.1 | 422 | 93 | |||
| XM_023643357.1 | XP_023499125.1 | 421 | 93 | |||
| XM_023643358.1 | XP_023499126.1 | 421 | 93 | |||
| XM_023643359.1 | XP_023499127.1 | 421 | 93 | |||
| XM_023643360.1 | XP_023499128.1 | 422 | 93 | |||
| XM_023643361.1 | XP_023499129.1 | 422 | 93 | |||
| XM_023643362.1 | XP_023499130.1 | 422 | 93 | |||
| XM_023643364.1 | XP_023499132.1 | 422 | 93 | |||
| XM_023643365.1 | XP_023499133.1 | 422 | 93 | |||
| XM_023643366.1 | XP_023499134.1 | 421 | 93 | |||
| XM_023643367.1 | XP_023499135.1 | 421 | 93 | |||
Set Membership: |
|