Search
Search

MCRS1| Gene Symbol: MCRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504207.5 | XP_001504257.1 | 792 | 67 | ||
| XM_001504209.3 | XP_001504259.1 | 869 | 67 | |||
| XM_023643509.1 | XP_023499277.1 | 798 | 67 | |||
| XM_023643511.1 | XP_023499279.1 | 883 | 67 | |||
Set Membership: |
|