Search
Search

PLCG1| Gene Symbol: PLCG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604632.3 | XP_005604689.1 | 897 | 108 | ||
| XM_023626566.1 | XP_023482334.1 | 1130 | 108 | |||
| XM_023626567.1 | XP_023482335.1 | 1202 | 108 | |||
Set Membership: |
|