Search
Search

RNF20| Gene Symbol: RNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504005.5 | XP_001504055.1 | 1190 | 61 | ||
| XM_005605666.3 | XP_005605723.1 | 1271 | 61 | |||
| XM_014735936.2 | XP_014591422.1 | 1165 | 61 | |||
| XM_023629772.1 | XP_023485540.1 | 1068 | 61 | |||
Set Membership: |
|