Search
Search

PRMT2| Gene Symbol: PRMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630373.1 | XP_023486141.1 | 552 | 70 | ||
| XM_023630374.1 | XP_023486142.1 | 845 | 70 | |||
| XM_023630375.1 | XP_023486143.1 | 410 | 70 | |||
| XM_023630376.1 | XP_023486144.1 | 556 | 70 | |||
| XM_023630377.1 | XP_023486145.1 | 549 | 70 | |||
| XM_023630378.1 | XP_023486146.1 | 841 | 70 | |||
| XM_023630380.1 | XP_023486148.1 | 552 | 70 | |||
Set Membership: |
|