Search
Search

MCPH1| Gene Symbol: MCPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495074.6 | XP_001495124.4 | 678 | 116 | ||
| XM_014736654.2 | XP_014592140.2 | 679 | 116 | |||
| XM_023630479.1 | XP_023486247.1 | 558 | 116 | |||
| XM_023630480.1 | XP_023486248.1 | 558 | 116 | |||
| XM_023630481.1 | XP_023486249.1 | 558 | 116 | |||
| XM_023630483.1 | XP_023486251.1 | 558 | 116 | |||
| XM_023630484.1 | XP_023486252.1 | 558 | 116 | |||
| XM_023630485.1 | XP_023486253.1 | 562 | 116 | |||
| XM_023630486.1 | XP_023486254.1 | 586 | 116 | |||
Set Membership: |
|