Search
Search

ZNF596| Gene Symbol: ZNF596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023630920.1 | XP_023486688.1 | 200 | 63 | ||
| XM_023630921.1 | XP_023486689.1 | 589 | 63 | |||
| XM_023630922.1 | XP_023486690.1 | 131 | 63 | |||
| XM_023630923.1 | XP_023486691.1 | 2407 | 63 | |||
| XM_023630927.1 | XP_023486695.1 | 2941 | 63 | |||
Set Membership: |
|