Search
Search

SNTB2| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023637203.1 | XP_023492971.1 | 1201 | 74 | ||
| XM_023637204.1 | XP_023492972.1 | 1205 | 74 | |||
| GenBank mRNA | JL635625.1 | - | 1 - 2 | 39 | 74 | |
Set Membership: |
|