Search
Search

CSNK1G2| Gene Symbol: CSNK1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023644337.1 | XP_023500105.1 | 1235 | 69 | ||
| XM_023644338.1 | XP_023500106.1 | 1195 | 69 | |||
| XM_023644339.1 | XP_023500107.1 | 1235 | 69 | |||
| XM_023644340.1 | XP_023500108.1 | 887 | 69 | |||
Set Membership: |
|