Search
Search

NFXL1| Gene Symbol: NFXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638133.1 | XP_023493901.1 | 2443 | 96 | ||
| XM_023638134.1 | XP_023493902.1 | 2420 | 96 | |||
| XM_023638135.1 | XP_023493903.1 | 2440 | 96 | |||
| XM_023638137.1 | XP_023493905.1 | 2417 | 96 | |||
Set Membership: |
|