Search
Search
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638371.1 | XP_023494139.1 | 300 | 70 | ||
| XM_023638372.1 | XP_023494140.1 | 300 | 70 | |||
| XM_023638373.1 | XP_023494141.1 | 315 | 70 | |||
| XM_023638374.1 | XP_023494142.1 | 300 | 70 | |||
| XM_023638375.1 | XP_023494143.1 | 300 | 70 | |||
| XM_023638376.1 | XP_023494144.1 | 300 | 70 | |||
Set Membership: |
|