Search
Search

PLAGL1| Gene Symbol: PLAGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023632983.1 | XP_023488751.1 | 834 | 80 | ||
| XM_023632984.1 | XP_023488752.1 | 5438 | 80 | |||
| XM_023632985.1 | XP_023488753.1 | 5544 | 80 | |||
| XM_023632986.1 | XP_023488754.1 | 5307 | 80 | |||
| XM_023632987.1 | XP_023488755.1 | 4300 | 80 | |||
Set Membership: |
|