Search
Search

SEMA3C| Gene Symbol: SEMA3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023638816.1 | XP_023494584.1 | 1410 | 68 | ||
| XM_023638817.1 | XP_023494585.1 | 1721 | 68 | |||
| XM_023638818.1 | XP_023494586.1 | 1525 | 68 | |||
Set Membership: |
|