Search
Search

PHF14| Gene Symbol: PHF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023639285.1 | XP_023495053.1 | 2665 | 90 | ||
| XM_023639286.1 | XP_023495054.1 | 2665 | 90 | |||
| XM_023639287.1 | XP_023495055.1 | 2665 | 90 | |||
| XM_023639288.1 | XP_023495056.1 | 2665 | 90 | |||
| XM_023639289.1 | XP_023495057.1 | 2666 | 90 | |||
| XM_023639290.1 | XP_023495058.1 | 2666 | 90 | |||
| XM_023639291.1 | XP_023495059.1 | 2666 | 90 | |||
Set Membership: |
|