Search
Search
| Gene Symbol: MSI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647029.1 | XP_023502797.1 | 782 | 67 | ||
| XM_023647030.1 | XP_023502798.1 | 782 | 67 | |||
| XM_023647031.1 | XP_023502799.1 | 782 | 67 | |||
| XM_023647032.1 | XP_023502800.1 | 626 | 67 | |||
| XM_023647033.1 | XP_023502801.1 | 782 | 67 | |||
Set Membership: |
|