Search
Search

MAPKAPK5| Gene Symbol: MAPKAPK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005612593.3 | XP_005612650.1 | 1012 | 91 | ||
| XM_005612594.3 | XP_005612651.1 | 1008 | 91 | |||
| XM_023648150.1 | XP_023503918.1 | 930 | 91 | |||
| XM_023648151.1 | XP_023503919.1 | 808 | 91 | |||
Set Membership: |
|