Search
Search

PPP4R1| Gene Symbol: PPP4R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023647459.1 | XP_023503227.1 | 2234 | 60 | ||
| XM_023647460.1 | XP_023503228.1 | 2183 | 60 | |||
| XM_023647461.1 | XP_023503229.1 | 2180 | 60 | |||
| XM_023647462.1 | XP_023503230.1 | 2129 | 60 | |||
Set Membership: |
|