Search
Search

CEP192| Gene Symbol: CEP192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014727718.2 | XP_014583204.2 | 6143 | 70 | ||
| XM_014727719.2 | XP_014583205.2 | 6143 | 70 | |||
| XM_014727721.2 | XP_014583207.2 | 6143 | 70 | |||
| XM_023647476.1 | XP_023503244.1 | 6143 | 70 | |||
| XM_023647477.1 | XP_023503245.1 | 5475 | 70 | |||
| XM_023647478.1 | XP_023503246.1 | 4963 | 70 | |||
| XM_023647479.1 | XP_023503247.1 | 4914 | 70 | |||
Set Membership: |
|