Search
Search

DEPTOR| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496931.6 | XP_001496981.4 | 582 | 65 | ||
| XM_023648901.1 | XP_023504669.1 | 582 | 65 | |||
| XM_023648902.1 | XP_023504670.1 | 1148 | 65 | |||
Set Membership: |
|