Search
Search

FAM91A1| Gene Symbol: FAM91A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497901.5 | XP_001497951.1 | 1446 | 73 | ||
| XM_023648925.1 | XP_023504693.1 | 1231 | 73 | |||
| XM_023648926.1 | XP_023504694.1 | 961 | 73 | |||