Search
Search

GNL3L| Gene Symbol: GNL3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495943.5 | XP_001495993.2 | 1486 | 69 | ||
| XM_005613881.3 | XP_005613938.1 | 1486 | 69 | |||
| XM_014728661.2 | XP_014584147.1 | 1458 | 69 | |||
Set Membership: |
|