Search
Search

SHROOM2| Gene Symbol: SHROOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023634059.1 | XP_023489827.1 | 566 | 80 | ||
| XM_023634060.1 | XP_023489828.1 | 567 | 80 | |||
| XM_023634061.1 | XP_023489829.1 | 1852 | 80 | |||
| XM_023634062.1 | XP_023489830.1 | 566 | 80 | |||
| XM_023634063.1 | XP_023489831.1 | 2138 | 80 | |||
Set Membership: |
|