Search
Search

FRMPD4| Gene Symbol: FRMPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023633847.1 | XP_023489615.1 | 1632 | 111 | ||
| XM_023633848.1 | XP_023489616.1 | 974 | 111 | |||
| XM_023633850.1 | XP_023489618.1 | 484 | 111 | |||
| XM_023633851.1 | XP_023489619.1 | 1629 | 111 | |||
Set Membership: |
|