Search
Search
| Gene Symbol: AMOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005614426.3 | XP_005614483.1 | 3192 | 67 | ||
| XM_005614427.3 | XP_005614484.1 | 2776 | 67 | |||
| XM_005614428.3 | XP_005614485.1 | 3043 | 67 | |||
| XM_023634126.1 | XP_023489894.1 | 2626 | 67 | |||
| XM_023634127.1 | XP_023489895.1 | 2108 | 67 | |||
| XM_023634128.1 | XP_023489896.1 | 1757 | 67 | |||
Set Membership: |
|