Search
Search

SMARCA1| Gene Symbol: SMARCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500518.6 | XP_001500568.1 | 2118 | 92 | ||
| XM_003365875.4 | XP_003365923.1 | 2082 | 92 | |||
| XM_005614487.3 | XP_005614544.1 | 2118 | 92 | |||
| XM_005614488.3 | XP_005614545.1 | 2118 | 92 | |||
| XM_014729163.2 | XP_014584649.1 | 2082 | 92 | |||
| XM_014729164.2 | XP_014584650.1 | 2118 | 92 | |||
Set Membership: |
|