Search
Search

BCORL1| Gene Symbol: BCORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014729166.2 | XP_014584652.2 | 4581 | 61 | ||
| XM_023634232.1 | XP_023490000.1 | 4793 | 61 | |||
| XM_023634233.1 | XP_023490001.1 | 4825 | 61 | |||
| XM_023634235.1 | XP_023490003.1 | 4553 | 61 | |||
| XM_023634236.1 | XP_023490004.1 | 5091 | 61 | |||
| XM_023634237.1 | XP_023490005.1 | 4376 | 61 | |||
| XM_023634238.1 | XP_023490006.1 | 4403 | 61 | |||
| XM_023634239.1 | XP_023490007.1 | 1513 | 61 | |||
Set Membership: |
|